Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | FARS2 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0941
![]()
|
Proteintech Group Inc
164361AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.Specifications
| FARS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O95363, Q6AYQ3, Q99M01 | |
| 10667, 306879, 69955 | |
| FARS2 Fusion Protein Ag9750 | |
| Primary | |
| -20°C | |
| FARS2 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| FARS1, FARS2, HSPC320, Phenylalanine tRNA ligase, PheRS | |
| Fars2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title