Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FARSA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528157 Supplier Invitrogen™ Supplier No. PA528157
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (93%), Rat (94%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARSA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene FARSA
Gene Accession No. Q8C0C7, Q9Y285
Gene Alias 0610012A19Rik; CML33; FARS; Farsa; FARSL; FARSLA; FRSA; PheHA; phenylalanine tRNA ligase 1, alpha, cytoplasmic; phenylalanine--tRNA ligase alpha chain; phenylalanine--tRNA ligase alpha subunit; phenylalanine-tRNA synthetase alpha-subunit; phenylalanine-tRNA synthetase-like, alpha subunit; phenylalanyl-tRNA synthetase alpha chain; phenylalanyl-tRNA synthetase alpha subunit; phenylalanyl-tRNA synthetase, alpha subunit; phenylalanyl-tRNA synthetase-like, alpha subunit; pheRS
Gene Symbols FARSA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FARSLA. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2193, 66590
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.