Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FARSA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112250 Supplier Invitrogen™ Supplier No. PA5112250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FARSA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARSA
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene FARSA
Gene Accession No. Q9Y285
Gene Alias 0610012A19Rik; CML33; FARS; Farsa; FARSL; FARSLA; FRSA; PheHA; phenylalanine tRNA ligase 1, alpha, cytoplasmic; phenylalanine--tRNA ligase alpha chain; phenylalanine--tRNA ligase alpha subunit; phenylalanine-tRNA synthetase alpha-subunit; phenylalanine-tRNA synthetase-like, alpha subunit; phenylalanyl-tRNA synthetase alpha chain; phenylalanyl-tRNA synthetase alpha subunit; phenylalanyl-tRNA synthetase, alpha subunit; phenylalanyl-tRNA synthetase-like, alpha subunit; pheRS
Gene Symbols FARSA
Host Species Rabbit
Immunogen E. coli-derived Human FARSA fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.