Learn More
Invitrogen™ FARSA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590609
Description
Immunogen sequence: YNFLAHGVLP DSGHLHPLLK VRSQFRQIFL EMGFTEMPTD NFIESSFWNF DALFQPQQHP ARDQHDTFFL RDPAEALQLP MDYVQRVKRT HSQGGYGSQG YKYNWKLDEA RKNLLRTHTT SASARALYRL AQKKPFTPVK YFSIDRVFRN ETLDATHLAE FHQIEGVVAD HGLTLGHLMG VLREFFTKLG ITQLRFKPAY NPYTEPSMEV FSYHQGLKKW VEVGNSGVFR PEMLLPMGLP ENVSVIAWGL SLERPTMIKY GINNIRELVG HKVNLQM; Positive Samples: HeLa, Mouse liver; Cellular Location: Cytoplasm.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
Specifications
FARSA | |
Polyclonal | |
Unconjugated | |
FARSA | |
0610012A19Rik; CML33; FARS; Farsa; FARSL; FARSLA; FRSA; PheHA; phenylalanine tRNA ligase 1, alpha, cytoplasmic; phenylalanine--tRNA ligase alpha chain; phenylalanine--tRNA ligase alpha subunit; phenylalanine-tRNA synthetase alpha-subunit; phenylalanine-tRNA synthetase-like, alpha subunit; phenylalanyl-tRNA synthetase alpha chain; phenylalanyl-tRNA synthetase alpha subunit; phenylalanyl-tRNA synthetase, alpha subunit; phenylalanyl-tRNA synthetase-like, alpha subunit; pheRS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2193, 66590 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.36 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8C0C7, Q9Y285 | |
FARSA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 210-486 of human FARSA (NP_0044521). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.