Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FARSB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120354 Supplier Invitrogen™ Supplier No. PA5120354
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: K-562, HeLa, Mouse kidney. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: DHSRITVNTR NIFIECTGTD FTKAKIVLDI IVTMFSEYCE NQFTVEAAEV VFPNGKSHTF PELAYRKEMV RADLINKKVG IRETPENLAK LLTRMYLKSE VIGDGNQIEI EIPPTRADII HACDIVEDAA IAYGYNNIQM TLPKTYTIAN QFPLNKLTEL LRHDMAAAGF TEALTFALCS QEDIADKLGV DISATKAVHI SNPKTAEFQV ARTTLLPGLL KTIAANRKMP LPLKLFEISD IVIKDSNTDV GAKNYRHLCA VYYNKNPGFE IIHGLLDRIM QLLDVPPGED KGGYVIKASE GPAFFPGRCA EIFARGQSVG KLGVLHPDVI TKFELTMPCS SLEINVGPFL.

This gene encodes a highly conserved enzyme that belongs to the aminoacyl-tRNA synthetase class IIc subfamily. This enzyme comprises the regulatory beta subunits that form a tetramer with two catalytic alpha subunits. In the presence of ATP, this tetramer is responsible for attaching L-phenylalanine to the terminal adenosine of the appropriate tRNA. A pseudogene located on chromosome 10 has been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARSB
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Farsb
Gene Accession No. Q9NSD9, Q9WUA2
Gene Alias C76708; Farsa; FARSB; Farsl; Farslb; Frsb; HSPC173; ph; PheHB; phenylalanine tRNA ligase 1, beta, cytoplasmic; phenylalanine tRNA synthetase alpha; phenylalanine-tRNA ligase beta chain; phenylalanine--tRNA ligase beta chain; phenylalanine--tRNA ligase beta subunit; phenylalanine-tRNA synthetase-like, beta subunit; phenylalanyl-tRNA synthetase beta chain; phenylalanyl-tRNA synthetase beta subunit; phenylalanyl-tRNA synthetase beta-subunit; phenylalanyl-tRNA synthetase, beta subunit; phenylalanyl-tRNA synthetase-like, beta subunit; PheRS; PheRS alpha
Gene Symbols Farsb
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-589 of human FARSB (NP_005678.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10056, 23874, 301544
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.