Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FARSB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563954 Supplier Invitrogen™ Supplier No. PA563954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IECTGTDFTK AKIVLDIIVT MFSEYCENQF TVEAAEVVFP NGKSHTFPEL AYRKEMVRAD LINKKVGIRE TPENLAKLLT RMYL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

This gene encodes a highly conserved enzyme that belongs to the aminoacyl-tRNA synthetase class IIc subfamily. This enzyme comprises the regulatory beta subunits that form a tetramer with two catalytic alpha subunits. In the presence of ATP, this tetramer is responsible for attaching L-phenylalanine to the terminal adenosine of the appropriate tRNA. A pseudogene located on chromosome 10 has been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARSB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Farsb
Gene Accession No. Q9NSD9
Gene Alias C76708; Farsa; FARSB; Farsl; Farslb; Frsb; HSPC173; ph; PheHB; phenylalanine tRNA ligase 1, beta, cytoplasmic; phenylalanine tRNA synthetase alpha; phenylalanine-tRNA ligase beta chain; phenylalanine--tRNA ligase beta chain; phenylalanine--tRNA ligase beta subunit; phenylalanine-tRNA synthetase-like, beta subunit; phenylalanyl-tRNA synthetase beta chain; phenylalanyl-tRNA synthetase beta subunit; phenylalanyl-tRNA synthetase beta-subunit; phenylalanyl-tRNA synthetase, beta subunit; phenylalanyl-tRNA synthetase-like, beta subunit; PheRS; PheRS alpha
Gene Symbols Farsb
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FARSB. Recombinant protein control fragment (Product #RP-103400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.