Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FARSLA/CML33 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982649
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2649 Supplier Bioss Supplier No. BS13145R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Functions as guanine nucleotide exchange factor for RAC1. May play a role in semaphorin signaling. Plays a role in the assembly and disassembly of dendritic filopodia, the formation of dendritic spines, regulation of dendrite length and ultimately the formation of synapses.

Specifications

Antigen FARSLA/CML33
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FARP1
Gene Accession No. Q9Y4F1
Gene Alias CDEP; Chondrocyte-derived ezrin-like protein; FARP1; FARP1-IT1; FERM, ARH/RhoGEF and pleckstrin domain protein 1; FERM, ARHGEF and pleckstrin domain-containing protein 1; FERM, RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived); FERM, RhoGEF and pleckstrin domain-containing protein 1; PH domain-containing family C member 2; Pleckstrin homology domain-containing family C member 2; PLEKHC2; PPP1R75; protein phosphatase 1, regulatory subunit 75
Gene Symbols FARP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FARSLA/CML33.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10160
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.