Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FARSLA Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FARSLA |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FARSLA Polyclonal specifically detects FARSLA in Human, Mouse samples. It is validated for Western Blot.Specifications
FARSLA | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
CML33pheRS, EC 6.1.1.20, FARS, FARSL, FARSLA, FRSA, PheHA, phenylalanine tRNA ligase 1, alpha, cytoplasmic, Phenylalanine--tRNA ligase alpha chain, phenylalanine-tRNA synthetase alpha-subunit, phenylalanine-tRNA synthetase-like, phenylalanine-tRNA synthetase-like, alpha subunit, phenylalanyl-tRNA synthetase alpha chain, phenylalanyl-tRNA synthetase, alpha subunit, phenylalanyl-tRNA synthetase-like, alpha subunit, PheRS | |
Recombinant fusion protein containing a sequence corresponding to amino acids 210-486 of human FARSLA (NP_004452.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
2193 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title