Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas Ligand/TNFSF6 Antibody (001), Biotin

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290737B
Description
Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Mouse samples. It is validated for ELISA.Specifications
Fas Ligand/TNFSF6 | |
Monoclonal | |
Biotin | |
apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fas Ligand/TNFSF6 (AAB02915.1; Pro132-Leu279). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Diabetes Research, Immunology, Inhibitors Activators and Regulators | |
356 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction