Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas Ligand/TNFSF6 Antibody (14C2), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063072APCCY7
Description
Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for Flow Cytometry.Specifications
Fas Ligand/TNFSF6 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
FASLG | |
Made to Human CD178 | |
0.1 mL | |
Primary | |
NB100-63072 recognizes the human CD178 cell surface antigen, a 40kDa glycoprotein also known as Fas ligand (CD95L). CD178 is a member of the TNF family, which is expressed by activated T lymphocytes and NK cells. The protein may exist as either a membrane bound or a cleaved soluble form. CD178 plays an important role in T cell development cytotoxicity. Binding of CD178 to Fas (CD95) results in the induction of apoptosis. Clone 14C2 is not reported to have a blocking function. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
14C2 | |
Flow Cytometry | |
apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Diabetes Research, Immunology, Inhibitors Activators and Regulators | |
356.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction