Learn More
Description
Specifications
Specifications
| Antigen | Fas Ligand/TNFSF6 |
| Applications | Western Blot, Immunoprecipitation, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 14C2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:25 - 1:50, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Western Blot (Negative) |
| Formulation | PBS (pH 7.4) with 0.09% Sodium Azide |
| Gene Accession No. | P48023 |
| Gene Alias | apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
