Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas/TNFRSF6/CD95 Antibody (APO-1-3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280082
Description
Fas Receptor/TNFRSF6/CD95 Monoclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional, CyTOF-ready.Specifications
Fas Receptor/TNFRSF6/CD95 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:5000, Flow Cytometry 1-2 ug/ml, Immunoprecipitation, Functional, CyTOF-ready | |
ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tumor necrosis receptor superfamily member 6, TNF receptor superfamily member 6, TNFRSF6, tumor necrosis factor receptor superfamily member 6, tumor necrosis factor receptor superfamily, member 6 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes human Fas Receptor/TNFRSF6/CD95. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG3 |
Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF | |
APO-1-3 | |
Unconjugated | |
PBS | |
FAS | |
Recombinant human Fas Receptor/TNFRSF6/CD95. | |
50 μg | |
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells | |
355 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction