Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas/TNFRSF6/CD95 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP233120
Description
Fas Receptor/TNFRSF6/CD95 Polyclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human, Primate samples. It is validated for Western Blot.Specifications
Fas Receptor/TNFRSF6/CD95 | |
Polyclonal | |
Unconjugated | |
10mM PBS - BSA free with No Preservative | |
FAS | |
Amino acids KDITSDSENSNFRNEIQSLV of human FAS (CD95) were used as the immunogen for this antibody. | |
0.1 mg | |
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells | |
355 | |
Human, Primate | |
Purified |
Western Blot | |
1.0 mg/mL | |
Western Blot 0.5-1ug/ml | |
ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tum | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
There are multiple isoforms of human FAS protein. Researchers are encouraged to use an online tool like NCBI BLAST to determine if this antibody is suitable for their use. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction