Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas/TNFRSF6/CD95 Antibody (B-D29), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314578AF350
Description
Fas/TNFRSF6/CD95 Monoclonal antibody specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for ELISASpecifications
Fas/TNFRSF6/CD95 | |
Monoclonal | |
Alexa Fluor 350 | |
Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6 | |
Recombinant Fas/TNFRSF6/CD95 | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells | |
355 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
B-D29 | |
50mM Sodium Borate | |
Mouse | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction