Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fas/TNFRSF6/CD95 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$207.00 - $501.50
Specifications
Antigen | Fas Receptor/TNFRSF6/CD95 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018071
![]() |
Novus Biologicals
NB120135500.025MG |
0.025 mg |
Each for $207.00
|
|
|||||
NB12013550
![]() |
Novus Biologicals
NB12013550 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
Fas Receptor/TNFRSF6/CD95 Polyclonal specifically detects Fas Receptor/TNFRSF6/CD95 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Fas Receptor/TNFRSF6/CD95 | |
Western Blot 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 | |
Polyclonal | |
Rabbit | |
Rat, Human, Mouse | |
P25446 | |
355 | |
Synthetic peptide: ESLKLRRRVHETDKNC conjugated to KLH, corresponding to amino acids 29-44 of Mouse CD95. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
45 kDa |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry | |
Unconjugated | |
RUO | |
PBS (pH 7.2) with 0.09% Sodium Azide | |
ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tum | |
FAS | |
IgG | |
Affinity Purified | |
CD95 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title