Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fascin 2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP106036
Description
Fascin 2 Polyclonal specifically detects Fascin 2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Fascin 2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotuspurpuratus), fascin-2, Retinal fascin, retinal), RFSNRP30fascin (Strongylocentrotus purpuratus) homolog 2 (actin-bundling protein | |
Goat | |
Affinity Purified | |
RUO | |
25794 | |
Mouse | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin 4 ug/mL, Peptide ELISA Detection limit 1:64000 | |
NP_001070650.1 | |
FSCN2 | |
Peptide with sequence CHHRGSNQLDTNR corresponding to internal region according to NP_036550.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_001070650.1 and NP_036550.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction