Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fascin-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140274
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TSHVLSAYHM WTPRPALHVH VILYSPIHRC YARADPTMGR IWVDAAVPCL EECGFLLHFR DGCYHLETST HHFLSHVDRL FSQPS.
FSCN3 (Fascin Actin-Bundling Protein 3) is a Protein Coding gene. Among its related pathways areMAPK-Erk Pathway. Gene Ontology (GO) annotations related to this gene includeactin filament bindingandprotein binding, bridging. An important paralog of this gene isFSCN1.
Specifications
Fascin-3 | |
Polyclonal | |
Unconjugated | |
FSCN3 | |
fascin (testis); fascin 3; fascin actin-bundling protein 3; fascin actin-bundling protein 3, testicular; fascin homolog 3, actin-bundling protein, testicular; fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotus purpuratus); fascin-3; Fscn3; testicular secretory protein Li 18; testis fascin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29999 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQT6 | |
FSCN3 | |
Recombinant protein corresponding to Human FSCN3. Recombinant protein control fragment (Product #RP-109457). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction