Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Fascin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1112S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1112S1 500 μL Concentrated
MS1112S0 100 μL Concentrated
MS1112S 1 mL Concentrated
MS1112R1 1 mL Prediluted
MS1112R7 7 mL Prediluted
MS1112RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1112S1 Supplier Epredia™ Supplier No. MS1112S1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Fascin Ab-1, Mouse Monoclonal Antibody.

Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predominantly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However, Reed Sternberg cells in Hodgkin's lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.

Host Species: Mouse

Clone: FCN01; same as 55K-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Fascin purified from HeLa cells

Molecular Weight: 55 kDa

Positive Control: Hodgkin's lymphoma or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Fascin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FCN01; same as 55K-2
Conjugate Unconjugated
Host Species Mouse
Immunogen Fascin purified from HeLa cells
Purification Method Hodgkin's lymphoma or spleen
Quantity 500 μL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.