Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fascin Antibody (FSCN1/417), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen Fascin
Clone FSCN1/417
Concentration 0.2mg/mL
Dilution Western Blot 0.5 - 1.0 ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24438300
SDP
View Documents
Novus Biologicals
NBP2443830.02MG
0.02 mg
Each for $268.00
Only null left
Add to Cart
 
NBP24438301
SDP
View Documents
Novus Biologicals
NBP2443830.1MG
0.1 mg
Each for $524.00
Only null left
Add to Cart
 
NBP24438302
SDP
View Documents
Novus Biologicals
NBP2443830.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Fascin Monoclonal specifically detects Fascin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Fascin
0.2mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Unconjugated
Mouse
Cytoskeleton Markers
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin
FSCN1
IgG2a κ
Protein A or G purified
Recognizes a protein of 55kDa, which is identified as fascin-1. Its actin binding ability is regulated by phosphorylation. Antibody to fascin-1 is a very sensitive marker for Reed-Sternberg cells and variants in nodular sclerosis, mixed cellularity, and lymphocyte depletion Hodgkin's disease. It is uniformly negative in lymphoid cells, plasma cells, and myeloid cells. Fascin-1 is also expressed in dendritic cells. This marker may be helpful to distinguish between Hodgkin lymphoma and non-Hodgkin lymphoma in difficult cases. Also, the lack of expression of fascin-1 in the neoplastic follicles in follicular lymphoma may be helpful in distinguishing these lymphomas from reactive follicular hyperplasia in which the number of follicular dendritic cells is normal or increased. Antibody to fascin-1 has been suggested as a prognostic marker in neuroendocrine neoplasms of the lung as well as in ovarian cancer. Fascin-1 expression may be induced by Epstein-Barr virus (EBV) infection of B cells with the possibility that viral induction of fascin in lymphoid or other cell types must also be considered in EBV-positive cases.
FSCN1/417
Western Blot 0.5 - 1.0 ug/ml, Simple Western 10 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human
Q16658
6624
Full length recombinant human FSCN1 protein
Primary
Store at 4C.
55 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.