Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fascin Antibody (OTI3B4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271294H
Description
Fascin Monoclonal specifically detects Fascin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Fascin | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| FSCN1 | |
| Full length recombinant protein of human Fascin (NP_003079) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3B4 | |
| Western Blot, Immunohistochemistry | |
| FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin | |
| Mouse | |
| Affinity Purified | |
| Cytoskeleton Markers | |
| 6624.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction