Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fascin Monoclonal Antibody (55K2)
GREENER_CHOICE

Catalog No. PIMA547716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547716 Supplier Invitrogen™ Supplier No. MA547716
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 55 kDa* band corresponding to fascin on SDS-PAGE immunoblots of human A431 and HeLa, as well as mouse C2C12 and rat PC12.

Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fascin
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 55K2
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene FSCN1
Gene Accession No. P85845, Q16658, Q61553
Gene Alias 55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658}
Gene Symbols FSCN1
Host Species Mouse
Immunogen Clone (55K2) was generated from full-length human fascin purified from HeLa cells. This sequence has high homology to rat and mouse fascin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14086, 6624, 683788
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.