Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fascin Mouse anti-Human, FITC, Clone: 13D2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259435F
This item is not returnable.
View return policy
Description
Fascin Monoclonal antibody specifically detects Fascin in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Fascin | |
Monoclonal | |
FITC | |
FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin | |
Recombinant human Fascin (1-493aa) purified from E. coli | |
0.1 mL | |
Cell Biology, Cellular Markers | |
6624 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry, ELISA, Immunofluorescence | |
13D2 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction