Learn More
Invitrogen™ Fascin Polyclonal Antibody

Description
Immunogen sequence: GEHGFIGCRK VTGTLDANRS SYDVFQLEFN DGAYNIKDST GKYWTVGSDS AVTSSGDTPV DFFFEFCDYN KVAIKVGGRY LKGDHAGVLK ASAETVDP Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 94%.
Specifications
Specifications
| Antigen | Fascin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FSCN1 |
| Gene Accession No. | Q16658 |
| Gene Alias | 55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658} |
| Gene Symbols | FSCN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.