Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fascin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6848R

Description
Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.Specifications
Fascin | |
Polyclonal | |
Unconjugated | |
FSCN1 | |
55 kDa actin-bundling protein; AI663989; Fan1; Fascin; fascin 1; fascin actin-bundling protein 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; singed-like (fascin homolog, sea urchin); Singed-like protein; SNL; UniProtKB:Q16658} | |
Rabbit | |
Protein A | |
RUO | |
6624 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q16658 | |
FSCN1 | |
KLH conjugated synthetic peptide derived from human Fascin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction