Learn More
Invitrogen™ Fascin Recombinant Rabbit Monoclonal Antibody (SAIC-32C-205)

Description
Specificity: Recognises human FSCN1.
Specifications
Specifications
| Antigen | Fascin |
| Applications | Flow Cytometry, Immunocytochemistry, Mass Spectrometry |
| Classification | Recombinant Monoclonal |
| Clone | SAIC-32C-205 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | FSCN1 |
| Gene Accession No. | Q16658 |
| Gene Alias | 55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658} |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.