Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FASN/Fatty Acid Synthase Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1498R

Description
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.Specifications
FASN/Fatty Acid Synthase | |
Polyclonal | |
Unconjugated | |
FASN | |
[Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; A630082H08Rik; BOS_19336; Enoyl-[acyl-carrier-protein] reductase; FAS; FASN; Fatty acid synthase; MGC14367; MGC15706; OA-519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; short chain dehydrogenase/reductase family 27X, member 1; thioesterase | |
Rabbit | |
Protein A | |
RUO | |
2194 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P49327 | |
FASN | |
KLH conjugated synthetic peptide derived from mouse FASN. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction