Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FASN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579238
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579238 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579238 Supplier Invitrogen™ Supplier No. PA579238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HepG2 whole cell, human hepatocellular carcinoma tumor tissue (HCCT), human 293T whole cell, human MCF-7 whole cell, human RT4 whole cell, human A549 whole cell, rat liver tissue, mouse lung tissue, mouse liver tissue, mouse NIH/3T3 whole cell. IHC: human intestinal cancer tissue. ICC/IF: T47D cell.

Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene FASN
Gene Accession No. P12785, P19096, P49327
Gene Alias [Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; A630082H08Rik; Enoyl-[acyl-carrier-protein] reductase; FAS; FASN; Fatty acid synthase; fatty acid synthase (EC 3.2.1.23); MGC14367; MGC15706; OA-519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; short chain dehydrogenase/reductase family 27X, member 1; thioesterase
Gene Symbols FASN
Host Species Rabbit
Immunogen E. coli-derived human FASN recombinant protein (Position: M1-E226).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14104, 2194, 50671
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.