Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FASN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA132355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA132355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA132355 Supplier Invitrogen™ Supplier No. PA132355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-32355 detects Fatty Acid Synthase from mouse, porcine, human, monkey and rat samples.

Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASN
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene FASN
Gene Accession No. P12276, P12785, P19096, P49327
Gene Alias [Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; A630082H08Rik; Enoyl-[acyl-carrier-protein] reductase; FAS; FASN; Fatty acid synthase; fatty acid synthase (EC 3.2.1.23); MGC14367; MGC15706; OA-519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; short chain dehydrogenase/reductase family 27X, member 1; thioesterase
Gene Symbols FASN
Host Species Rabbit
Immunogen A synthetic peptide, conjugated to KLH, made near the N-terminus of mouse FAS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100689207, 14104, 2194, 396061, 397561, 50671
Target Species Human, Mouse, Rat, Monkey, Hamster, Pig, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.