Learn More
FASN Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00305T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Goat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Monkey, Gorilla, Chimpanzee and h
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.Specifications
FASN | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P19096, P49327 | |
FASN | |
Between 2462 and 2511 | |
10 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
FASN | |
FAS, MGC14367, MGC15706, OA-519, SDR27X1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14104, 2194 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.