Learn More
Epredia™ Fast Flex™ Holding Solution
Catalog No. p-4532395
Click to view available options
Quantity:
5L
2.5 gal.
Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.
  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives
Specifications
Quantity 2.5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.