Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fast Skeletal Myosin Heavy Chain 1 Antibody (LM5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250401B
Description
Fast Skeletal Myosin Heavy Chain 1 Monoclonal specifically detects Fast Skeletal Myosin Heavy Chain 1 in Chicken samples. It is validated for Western Blot.Specifications
Fast Skeletal Myosin Heavy Chain 1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Purified chicken pectorial muscle | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
LM5 | |
Western Blot | |
Mouse | |
Protein A purified | |
RUO | |
Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction