Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fast Skeletal Myosin Heavy Chain 1 Antibody (LM5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
| Antigen | Fast Skeletal Myosin Heavy Chain 1 |
|---|---|
| Clone | LM5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000 |
| Applications | Western Blot |
Description
Fast Skeletal Myosin Heavy Chain 1 Monoclonal antibody specifically detects Fast Skeletal Myosin Heavy Chain 1 in Chicken samples. It is validated for Western BlotSpecifications
| Fast Skeletal Myosin Heavy Chain 1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chicken | |
| Purified chicken pectorial muscle | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| LM5 | |
| Western Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites | |
| IgG | |
| Ascites |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title