Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fast skeletal myosin light chain 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216433
Description
Fast skeletal myosin light chain 1 Polyclonal specifically detects Fast skeletal myosin light chain 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Fast skeletal myosin light chain 1 | |
Polyclonal | |
Unconjugated | |
PBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
A1 catalytic, A2 catalytic, MLC1/MLC3, MLC1F, MLC1F/MLC3F, MLC3F, myosin light chain 1/3, skeletal muscle isoform, Myosin light chain A1/A2, Myosin light chain alkali 1/2, myosin, light chain 1, alkali; skeletal, fast, myosin, light polypeptide 1, alkali; skeletal, fast | |
Rabbit | |
17 kDa | |
0.1 mL | |
Cytoskeleton Markers, Vision | |
4632 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.59mg/mL | |
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
P06741 | |
MYL1 | |
Recombinant protein encompassing a sequence within the center region of human Fast skeletal myosin light chain 1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat. Expected to cross react based on sequence identity: Chicken (82%), Pig (94%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction