Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fast skeletal myosin light chain 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | Fast skeletal myosin light chain 1 |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Fast skeletal myosin light chain 1 Polyclonal antibody specifically detects Fast skeletal myosin light chain 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Fast skeletal myosin light chain 1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers, Vision | |
PBS, pH 7.2, 40% glycerol | |
4632 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
A1 catalytic, A2 catalytic, MLC1/MLC3, MLC1F, MLC1F/MLC3F, MLC3F, myosin light chain 1/3, skeletal muscle isoform, Myosin light chain A1/A2, Myosin light chain alkali 1/2, myosin, light chain 1, alkali; skeletal, fast, myosin, light polypeptide 1, alkali; skeletal, fast | |
This antibody was developed against Recombinant Protein corresponding to amino acids: APAPAKPKEEKIDLSAIKIEFSKEQQDEF | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title