Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Fast skeletal myosin light chain 1 Recombinant Protein Antigen
SDP

Catalog No. NB170967
Click to view available options
:
100 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 4632
Molecular Weight (g/mol) M.W. Theoretical: 21 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias A1 catalytic, A2 catalytic, MLC1/MLC3, MLC1F, MLC1F/MLC3F, MLC3F, myosin light chain 1/3, skeletal muscle isoform, Myosin light chain A1/A2, Myosin light chain alkali 1/2, myosin, light chain 1, alkali; skeletal, fast, myosin, light polypeptide 1, alkali; skeletal, fast
Gene Symbol MYL1
Research Category Cell Biology, Cellular Markers, Vision
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Fast skeletal myosin light chain 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.