Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast skeletal Myosin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986374
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6374 Supplier Bioss Supplier No. BS5159R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin is a 480 kD protein that interacts with actin in muscle and non muscle cells (8). Conventional myosin contains two identical heavy chains (MHC - 200kD each) and four light chains (MLC-15-26 kD). Conventional myosin molecules consist of two major regions: tail (rod) and globular heads; they aggregate into filaments through the tail region and interact with actin and with adenosine triphosphate (ATP) through the head region. Multiple forms of myosin heavy chains exist in skeletal, smooth and cardiac muscle and in non- muscle tissues. A spectrum of skeletal muscle fiber types is found in adult skeletal muscles. The two major skeletal muscle fiber types are type I (slow- twitch) and type II (fast-twitch). Skeletal muscle type II fibers can be further subdivided into types IIa (fast red) and IIb (fast white).

Specifications

Antigen Fast skeletal Myosin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MRLC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.