Learn More
Invitrogen™ FASTK Monoclonal Antibody (2B7)

Description
Peptide Sequence: KWGDRPVGGG PSAGPVQGLQ RLLEQAKSPG ELLRWLGQNP SKVRAHHYSV ALRRLGQLLG SRPRPPPVEQ VTLQDLSQLI IRNCPSFD.
Specifications
Specifications
| Antigen | FASTK |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2B7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FASTK |
| Gene Accession No. | Q14296 |
| Gene Alias | 0610011K02Rik; 610011K02Rik; Fas activated serine/threonine kinase; Fas-activated serine/threonine kinase; FAST; FAST kinase; Fastk |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.