Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FASTKD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549834
Description
The antibody detects endogenous levels of total FASTKD1 protein.
As a member of the FASTKD family, FASTKD1 localizes to the mitochondria to modulate their energy balance, especially under conditions of stress. Though ubiquitously expressed in all tissues, FASTKD1 appears more abundantly in skeletal muscle, heart muscle, and other tissues enriched in mitochondria. FASTKD1 has been validated as an RNA-binding protein.
Specifications
FASTKD1 | |
Polyclonal | |
Unconjugated | |
FASTKD1 | |
5330408N05Rik; FAST kinase domain-containing protein 1; FAST kinase domain-containing protein 1, mitochondrial; FAST kinase domains 1; Fastkd1; KIAA1800; mKIAA1800; RGD1563531 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
320720, 79675 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q53R41, Q6DI86 | |
FASTKD1 | |
Synthesized peptide derived from internal of human FAKD1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction