Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FASTKD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560483 Supplier Invitrogen™ Supplier No. PA560483
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSHNIALGQL PEMPWESNIE IVGSRLPPGA ERIALEFLDS KALCRNIPHM KGKSAMKKRH LEILGYRVIQ ISQFEWNSMA LSTKDA Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 64%.

As a member of the FASTKD family, FASTKD1 localizes to the mitochondria to modulate their energy balance, especially under conditions of stress. Though ubiquitously expressed in all tissues, FASTKD1 appears more abundantly in skeletal muscle, heart muscle, and other tissues enriched in mitochondria. FASTKD1 has been validated as an RNA-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASTKD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FASTKD1
Gene Accession No. Q53R41
Gene Alias 5330408N05Rik; FAST kinase domain-containing protein 1; FAST kinase domain-containing protein 1, mitochondrial; FAST kinase domains 1; Fastkd1; KIAA1800; mKIAA1800; RGD1563531
Gene Symbols FASTKD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FASTKD1. Recombinant protein control fragment (Product #RP-100809).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.