Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FASTKD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109711 Supplier Invitrogen™ Supplier No. PA5109711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFLARLDSQL EILSPSRSAR VQFHLMELNR SVCLECPEFQ IPWFHDRFCQ QYNKGIGGMD GTQQQIFKML AEVLGGINCV KASVLTPYYH KVDFECILDK RKKPLPYGSH NIALGQLPEM PWESNIEIVG SRLPPGAERI ALEFLDSKAL CRNIPHMKGK SAMKKRHLEI LGYRVIQISQ FEWNSMALST KDARMDYLRE CIFGEVKSCL.

As a member of the FASTKD family, FASTKD1 localizes to the mitochondria to modulate their energy balance, especially under conditions of stress. Though ubiquitously expressed in all tissues, FASTKD1 appears more abundantly in skeletal muscle, heart muscle, and other tissues enriched in mitochondria. FASTKD1 has been validated as an RNA-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASTKD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FASTKD1
Gene Accession No. Q53R41, Q6DI86
Gene Alias 5330408N05Rik; FAST kinase domain-containing protein 1; FAST kinase domain-containing protein 1, mitochondrial; FAST kinase domains 1; Fastkd1; KIAA1800; mKIAA1800; RGD1563531
Gene Symbols FASTKD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 638-847 of human FASTKD1 (NP_078898.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311112, 320720, 79675
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.