Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FASTKD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $727.54
Specifications
| Antigen | FASTKD2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FASTKD2 Polyclonal specifically detects FASTKD2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| FASTKD2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 22868 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QQVWKIEDVFTLQVVMKCIGKDAPIALKRKLEMKALRELDRFSVLNSQHMFEVLAAMNHRSLILLDECSKVVLDNIHGCPLRIMINILQSCKDLQY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| FAST kinase domains 2, KIAA0971FAST kinase domain-containing protein 2 | |
| FASTKD2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title