Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FASTKD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113444
Description
FASTKD2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency.
Specifications
FASTKD2 | |
Polyclonal | |
Unconjugated | |
Fastkd2 | |
2810421I24Rik; FAST kinase domain-containing protein 2; FAST kinase domain-containing protein 2, mitochondrial; FAST kinase domains 2; Fastkd2; KIAA0971; RGD1307883 | |
Rabbit | |
Protein G | |
RUO | |
22868, 301463, 75619 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4.09 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q5M7V7, Q922E6, Q9NYY8 | |
Fastkd2 | |
Recombinant Human FAST kinase domain-containing protein 2, mitochondrial protein (117-203AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction