Learn More
FASTKD2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303788AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency. [provided by RefSeq, Oct 2008].Specifications
FASTKD2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NYY8 | |
FASTKD2 | |
Between 660 and 710 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FASTKD2 | |
KIAA0971 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22868 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.