Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FASTKD3 Polyclonal Antibody
FASTKD3 Polyclonal Antibody
Supplier: Thermo Scientific PA550415
Description
FASTKD3 Polyclonal Antibody for Western Blot

Specifications
FASTKD3 | |
Polyclonal | |
Unconjugated | |
FASTKD3 | |
2310010B21Rik; AW048587; FAKD3; FAST kinase domain-containing protein 3; FAST kinase domain-containing protein 3, mitochondrial; FAST kinase domains 3; FASTKD3; FLJ23274; RGD1309729 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79072 | |
-20°C | |
Liquid |
Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q14CZ7 | |
FASTKD3 | |
Fusion protein corresponding to a region derived from internal residues of human FAST kinase domains 3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction