Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FASTKD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567348 Supplier Invitrogen™ Supplier No. PA567348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: METLPDTMAAG ALQRICEVEK KDGDQGLPKE ILENSIFQAL CFQFEKEPSQ LSNTSLVTAL QALILLHVDP QSSLLLNLVA ECQNRLRKGG MEVRNLCILG ESLITLHSSG CVTLELIINQ LQG Highest antigen sequence identity to the following orthologs - mouse 56%, rat 57%.

This gene encodes a member of a small family of Fas-activated serine/threonine kinase domain (FASTKD) containing proteins that share an amino terminal mitochondrial targeting domain and multiple carboxy terminal FAST domains as well as a putative RNA-binding RAP domain. The members of this family are ubiquitously expressed and are generally most abundant in mitochondria-enriched tissues such as heart, skeletal muscle and brown-adipose tissue. Some members of this protein family may play a role in apoptosis. The protein encoded by this gene interacts with components of the mitochondrial respiratory and translation networks. A pseudogene of this gene is also present on chromosome 5. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASTKD3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FASTKD3
Gene Accession No. Q14CZ7
Gene Alias 2310010B21Rik; AW048587; FAKD3; FAST kinase domain-containing protein 3; FAST kinase domain-containing protein 3, mitochondrial; FAST kinase domains 3; FASTKD3; FLJ23274; RGD1309729
Gene Symbols FASTKD3
Host Species Rabbit
Immunogen Recombinant Human FASTKD3. Recombinant protein control fragment (Product #RP-108000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.