Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FASTKD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115210 Supplier Invitrogen™ Supplier No. PA5115210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FAKD5.

Plays an important role in the processing of non-canonical mitochondrial mRNA precursors (PubMed:25683715). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASTKD5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FASTKD5
Gene Accession No. Q7L8L6, Q7TMV3
Gene Alias C78212; dJ1187M17.5; FAST kinase domain-containing protein 5; FAST kinase domain-containing protein 5, mitochondrial; FAST kinase domains 5; FAST kinase domains 5, pseudogene 1; Fastkd5; Fastkd5-ps1; KIAA1792; mKIAA1792; RGD1563839
Gene Symbols FASTKD5
Host Species Rabbit
Immunogen A synthesized peptide derived from human FASTKD5(Accession Q7L8L6), corresponding to amino acid residues N531-I581.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 380601, 60493
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.