Learn More
Invitrogen™ FASTKD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109712
Description
Immunogen sequence: SPGFVRLAQE RTKFDLLKEL YTLDGTVGIE CPDYRGNRLS THLQQEGSEL LWYLAEKDMN SKPEFLETVF LLETMLGGPQ YVKHHMILPH TRSSDLEVQL DVNLKPLPFN REATPAENVA KLRLEHVGVS LTDDLMNKLL KGKARGHFQG KTESEPGQQP MELENKAAVP LGGFLCNVAD KSGAMEMAGL CPAACMQTPR MKLAVQFTNR NQYCYGSRDL LGLHNMKRRQ LARLGYRVVE LSYWEWLPLL KRTRLEKLAF LHEKVFTSAL.
Plays an important role in the processing of non-canonical mitochondrial mRNA precursors (PubMed:25683715). [UniProt].
Specifications
FASTKD5 | |
Polyclonal | |
Unconjugated | |
FASTKD5 | |
C78212; dJ1187M17.5; FAST kinase domain-containing protein 5; FAST kinase domain-containing protein 5, mitochondrial; FAST kinase domains 5; FAST kinase domains 5, pseudogene 1; Fastkd5; Fastkd5-ps1; KIAA1792; mKIAA1792; RGD1563839 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
366192, 380601, 60493 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.74 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q7L8L6, Q7TMV3 | |
FASTKD5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 495-764 of human FASTKD5 (NP_068598.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.