Learn More
Restek™ FastPack™ Inlet Kits
SDP
Catalog No. 06611270 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Topaz Precision
Topaz Single Taper Splitless
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-270 Topaz Precision
06-611-269 Topaz Single Taper Splitless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-270 Supplier Restek™ Supplier No. 23259
Includes all necessary parts for maintenance such as replacing the injection port (inlet) liner, the critical inlet seals, and the septum
  • Injection port maintenance should be performed on a routine basis, and prior to installing any capillary column
  • FastPack Inlet Kit includes all necessary parts for maintenance such as replacing the injection port (inlet) liner, the critical inlet seals, and the septum
  • For optimum column performance, the injection port (inlet) liner must be free of septum particles, sample residue, or ferrule fragments
  • Peak shape degradation, poor reproducibility, sample decomposition, and ghost peaks all are associated with using a contaminated liner
  • The critical seal must fit tightly around the inlet liner to prevent carrier gas from leaking around the outside of the liner
  • Frequent septum replacement prevents fragmentation or leaks, and maintains a leak-tight seal to exclude air from the inlet
  • Thermolite Plus septa are preconditioned for quicker instrument uptime
  • Always use gloves and forceps when handling septa to avoid contamination
  • The only way to maintain optimum performance is by frequently changing the inlet seal and ensuring the seal is leak tight.
Specifications
Type Topaz Precision
Height (Metric) 11 mm
Length (Metric) 4 mm
Material Multiple
Packing Material Wool
For Use With (Application) Injection Port Maintenance
Quantity 5 Kits/Pk.
For Use With (Equipment) Agilent GCs