Learn More
Fasudil monohydrochloride, 99+%
Description
Fasudil monohydrochloride is used in the prevention and treating cardiovascular and cerebrovascular diseases. It is also used in the inhibition of proliferation of vascular smooth muscle cells. It serves as a selective Rho-associated protein kinase (ROCK) inhibitor. Further, it protects against methyl mercury-induced axonal degeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Incompatible with acids, bases and oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Merck Index | 14,3942 |
| Solubility Information | Soluble in water and dimethyl sulfoxide. Insoluble in ethanol. |
| Formula Weight | 327.83 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Chemical Name or Material | Fasudil monohydrochloride |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| RTECSNumber | HM4031166 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.