Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAT1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232275AF594
Description
FAT1 Polyclonal specifically detects FAT1 in Human samples. It is validated for Western Blot.Specifications
FAT1 | |
Polyclonal | |
Western Blot | |
FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) | |
Rabbit | |
100 μL | |
2195 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
FAT1 | |
The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195). | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction